Etyyli-3-fluoribentsoaatti ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L07984.09 L07984.18
L07984.09EA 45.4 EUR
L07984.09 L07984.18
Etyyli-3-fluoribentsoaatti ≥98%
Etyyli-3-fluoribentsoaatti

Purity: 98+% CAS No.: 451-02-5 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 82°(179°F) Refractive Index: 1.4850 Odour: Recognizable Boiling Point: 208° Beilstein Reference: 2045876 MDL No.: MFCD00000336

Kaava: C₉H₉FO₂
Kiehumispiste: 208 °C
Tiheys: 1,136
Leimahduspiste: 82 °C (179 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000336
CAS-numero: 451-02-5
EINECS: 207-191-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR