Ethyl-4-nitrophenylglyoxylate ≥98%

Toimittaja: Thermo Fisher Scientific

L07579.03 L07579.06
L07579.03EA 149 EUR
L07579.03 L07579.06
Ethyl-4-nitrophenylglyoxylate ≥98%
Ethyl-4-nitrophenylglyoxylate

Purity: 98+% CAS No.: 70091-75-7 Molecular Formula: C10H9NO5 Formula Weight: 223.19 Melting Point: 39-43° Boiling Point: 152-157°/0.5mm MDL No.: MFCD00051841

Kaava: C₁₀H₉NO₅
Kiehumispiste: 152…157 °C (0,5 mmHg)
Sulamispiste: 39…43 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051841
CAS-numero: 70091-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR