3-Butyn-2-oni ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

L05527.03 L05527.06 L05527.14
L05527.03EA 146 EUR
L05527.03 L05527.06 L05527.14
3-Butyn-2-oni ≥98%
3-Butyn-2-oni

Purity: 98% CAS No.: 1423-60-5 Molecular Formula: C4H4O Formula Weight: 68.08 Storage Temperature: 2°C to 8°C Flash Point: -1°(30°F) Refractive Index: 1.4060 Boiling Point: 84-86° Beilstein Reference: 605353 UN No.: 1992 MDL No.: MFCD00008775

Kaava: C₄H₄O
Kiehumispiste: 84…86 °C
Tiheys: 0,870
Leimahduspiste: –1 °C (30 °F)
MDL numero: MFCD00008775
CAS-numero: 1423-60-5
EINECS: 215-834-2
UN: 1992
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR