Ethyl-4-hydroxymandelate 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L05413.03 L05413.06
L05413.03EA 69.3 EUR
L05413.03 L05413.06
Ethyl-4-hydroxymandelate 98%
Ethyl-4-hydroxymandelate

Purity: 98% CAS No.: 68758-68-9 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 128-132° Beilstein Reference: 3132065 MDL No.: MFCD00020180

Kaava: C₁₀H₁₂O₄
Sulamispiste: 128…132 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00020180
CAS-numero: 68758-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR