(1R)-(+)-α-Pinene ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L04941.18 L04941.30
L04941.18EA 125 EUR
L04941.18 L04941.30
(1R)-(+)-α-Pinene ≥98%
(1R)-(+)-α-Pinene

Purity: 98% CAS No.: 7785-70-8 Molecular Formula: C10H16 Formula Weight: 136.24 Flash Point: 32°(89°F) Refractive Index: 1.4660 Melting Point: -62° Boiling Point: 154-156° Beilstein Reference: 3648264 Merck Reference: 14,7445 UN No.: 2368 MDL No.: MFCD00001346

Kaava: C₁₀H₁₆
Kiehumispiste: 154…156 °C
Sulamispiste: –62 °C
Tiheys: 0,858
Leimahduspiste: 32 °C (89 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001346
CAS-numero: 7785-70-8
EINECS: 232-087-8
UN: 2368
ADR: 3,III
Merck Index: 14,7445

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR