2-Amino-4'-bromoacetophenone hydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L04292.04 L04292.09
L04292.04EA 107 EUR
L04292.04 L04292.09
2-Amino-4'-bromoacetophenone hydrochloride 98%
2-Amino-4'-bromoacetophenone hydrochloride

Purity: 98% CAS No.: 5467-72-1 Molecular Formula: C8H8BrNO.HCl Formula Weight: 250.53 Melting Point: 280-283° Beilstein Reference: 3630035

Kaava: C₈H₉BrClNO
Sulamispiste: 280…283 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051998
CAS-numero: 5467-72-1
EINECS: 226-778-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR