exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L03620.14 L03620.22
L03620.14EA 136 EUR
L03620.14 L03620.22
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98%
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride

Purity: 98+% CAS No.: 6118-51-0 Molecular Formula: C8H6O4 Formula Weight: 166.13 Melting Point: 118-119° Beilstein Reference: 10355 MDL No.: MFCD00151506

Kaava: C₈H₆O₄
Sulamispiste: 118…119 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151506
CAS-numero: 6118-51-0
EINECS: 226-570-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR