(2-Syanoetyyli)trietoksisilaani ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L03543.14 L03543.22
L03543.14EA 112 EUR
L03543.14 L03543.22
(2-Syanoetyyli)trietoksisilaani ≥97%
(2-Syanoetyyli)trietoksisilaani

Purity: 97% CAS No.: 919-31-3 Molecular Formula: NCCH2CH2Si(OCH2CH3)3 Formula Weight: 217.34 Flash Point: 100°(212°F) Refractive Index: 1.4130 Boiling Point: 224-225° Beilstein Reference: 1776392 MDL No.: MFCD00019851

Kaava: NCCH₂CH₂Si(OCH₂CH₃)₃
Kiehumispiste: 224…225 °C
Tiheys: 0,980
Leimahduspiste: 100 °C (212 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00019851
CAS-numero: 919-31-3
EINECS: 213-050-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR