3-Pentyn-1-oli ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L02951.03 L02951.06
L02951.03EA 67.6 EUR
L02951.03 L02951.06
3-Pentyn-1-oli ≥98%
3-Pentyn-1-oli

Purity: 98% CAS No.: 10229-10-4 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 54°(129°F) Refractive Index: 1.4560 Odour: Recognizable Boiling Point: 154-157° Beilstein Reference: 1735967 UN No.: 1987 MDL No.: MFCD00002956

Kaava: C₅H₈O
Kiehumispiste: 154…157 °C
Tiheys: 0,912
Leimahduspiste: 54 °C (129 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002956
CAS-numero: 10229-10-4
EINECS: 233-550-7
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR