1,3-Difenyyliasetoni ≥98%

Toimittaja: Thermo Fisher Scientific

L02830.22 L02830.14 L02830.PG
L02830.22EA 392 EUR
L02830.22 L02830.14 L02830.PG
1,3-Difenyyliasetoni ≥98%
1,3-Difenyyliasetoni

Purity: 98+% CAS No.: 102-04-5 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 330° Beilstein Reference: 974767 MDL No.: MFCD00004795

Kaava: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
Kiehumispiste: 330 °C
Sulamispiste: 32…36 °C
Tiheys: 1,04
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004795
CAS-numero: 102-04-5
EINECS: 203-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR