Tetrabutylammonium hydroxide 40% (w/w) vesiliuos

Toimittaja: Thermo Fisher Scientific
Vaara

L02809.18 L02809.30 L02809.0B
L02809.18EA 97.3 EUR
L02809.18 L02809.30 L02809.0B
Tetrabutylammonium hydroxide 40% (w/w) vesiliuos
Tetrabutylammonium hydroxide

CAS No.: 2052-49-5 Molecular Formula: C16H37NO Formula Weight: 259.48 Flash Point: None Refractive Index: 1.4080 Odour: Amine-like pH: 14.0 Beilstein Reference: 3571228 UN No.: 3267 MDL No.: MFCD00009425

Kaava: C₁₆H₃₇NO
Tiheys: 0,990
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009425
CAS-numero: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR