Lucifer yellow CH dilithium salt for fluorescence Stain, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ100

L0259-100MG L0259-250MG L0259-25MG
L0259-100MGEA 736 EUR
L0259-100MG L0259-250MG L0259-25MG
Lucifer yellow CH dilithium salt for fluorescence Stain, Sigma-Aldrich®
Lucifer yellow CH dilithium salt

A highly fluorescent dye, useful in marking nerve cells.

Kaava: C₁₃H₉N₅O₉S₂Li₂
Storage Temperature: Jääkaappi
MDL numero: MFCD00006922
CAS-numero: 67769-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0d89ade9323ed5759e602fcb51f539c612cb67e&vid=cb0c5c90571f8f53ea8b8cc60bd2700b1d7aa63f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance (Colour) Faint Orange to Very Dark Orange
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour)
1 mg/ml H2O
Faint Yellow to Very Dark Yellow and Faint Orange to Very Dark Orange
Solubility (Turbidity) Clear to Slightly Hazy
Fluorescence Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR