Dibenzyl ketoxime ≥98%

Toimittaja: Thermo Fisher Scientific

L00914.06 L00914.14
L00914.06EA 85.4 EUR
L00914.06 L00914.14
Dibenzyl ketoxime ≥98%
Dibenzyl ketoxime

Purity: 98+% CAS No.: 1788-31-4 Molecular Formula: C15H15NO Formula Weight: 225.29 Melting Point: 122-124° Beilstein Reference: 1965977 MDL No.: MFCD00015453

Kaava: C₁₅H₁₅NO
Sulamispiste: 122…124 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015453
CAS-numero: 1788-31-4
EINECS: 217-251-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR