Ethyl diethoxyacetate ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L00390.14 L00390.22
L00390.14EA 112 EUR
L00390.14 L00390.22
Ethyl diethoxyacetate ≥97%
Ethyl diethoxyacetate

Purity: 97+% CAS No.: 6065-82-3 Molecular Formula: C8H16O4 Formula Weight: 176.21 Flash Point: 85°(185°F) Refractive Index: 1.4100 Boiling Point: 198-200° Beilstein Reference: 878579 MDL No.: MFCD00009231

Kaava: C₈H₁₆O₄
Kiehumispiste: 198…200 °C
Tiheys: 0,988
Leimahduspiste: 85 °C (185 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009231
CAS-numero: 6065-82-3
EINECS: 227-999-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR