1,8-Bis(dimetyyliamino)naftaleeni ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

L00313.09 L00313.18
L00313.09EA 108 EUR
L00313.09 L00313.18
1,8-Bis(dimetyyliamino)naftaleeni ≥98%
1,8-Bis(dimetyyliamino)naftaleeni

Purity: 98+% CAS No.: 20734-58-1 Molecular Formula: C14H18N2 Formula Weight: 214.31 Flash Point: 110°(230°F) Melting Point: 47-51° Boiling Point: 144-145°/7mm Beilstein Reference: 396782 UN No.: 3263 MDL No.: MFCD00003920

Kaava: C₁₄H₁₈N₂
Kiehumispiste: 144…145 °C (7 mmHg)
Sulamispiste: 47…51 °C
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003920
CAS-numero: 20734-58-1
EINECS: 244-001-6
UN: 3263
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR