Azidotrimethylsilane ≥94%

Toimittaja: Thermo Fisher Scientific
Vaara

L00173.06 L00173.14 L00173.22
L00173.06EA 72.2 EUR
L00173.06 L00173.14 L00173.22
Azidotrimethylsilane ≥94%
Azidotrimethylsilane

Purity: 94% CAS No.: 4648-54-8 Molecular Formula: C3H9N3Si Formula Weight: 115.21 Storage Temperature: 2°C to 8°C Flash Point: 6°(42°F) Refractive Index: 1.4140 Melting Point: -95° Boiling Point: 95-99° Beilstein Reference: 1903730 UN No.: 1992 MDL No.: MFCD00001986

Kaava: C₃H₉N₃Si
Kiehumispiste: 95…99 °C
Sulamispiste: –95 °C
Tiheys: 0,872
Leimahduspiste: 6 °C (42 °F)
MDL numero: MFCD00001986
CAS-numero: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: 3,II

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR