Pharmacy fume hoods, benchtop
Toimittaja: KOETTERMANN
451-00011 451-00000 451-00001 451-00002 451-00003 451-00004 451-00005 451-00006 451-00007 451-00008 451-00010 451-00009
KOTT451-00011EA 0 EUR
KOTT451-00011 KOTT451-00000 KOTT451-00001 KOTT451-00002 KOTT451-00003 KOTT451-00004 KOTT451-00005 KOTT451-00006 KOTT451-00007 KOTT451-00008 KOTT451-00010 KOTT451-00009
Pharmacy fume hoods, benchtop
Vetokaapit ja kotelot Vetolääkekaapit
The pharmacy hood is a small hood, which was developed especially for the handling of hazardous substances in pharmacies. It complies with the requirements of section 4 (2) of the German Pharmacies Act and is ideally suited for handling dusty compounds or volatile chemicals. These hoods have been designed for safe handling of chemicals, mineral acids, concentrated and heated acids.
  • Easily mounted on a table top
  • Resistant to chemicals
  • Sash glazed safety glass
  • Workplace lighting
Construction: Epoxy powder coated steel housing, interior SGL lining.