Chemical storage cabinet
Toimittaja: KOETTERMANN
375-00015 375-00016 375-00017 375-00018 375-00019 375-00020 375-00012 375-00014 375-00001 375-00002 375-00003 375-00004 375-00005 375-00006 375-00007 375-00008 375-00009 375-00010 375-00011 375-00013
KOTT375-00015EA 3120 EUR
KOTT375-00015 KOTT375-00016 KOTT375-00017 KOTT375-00018 KOTT375-00019 KOTT375-00020 KOTT375-00012 KOTT375-00014 KOTT375-00001 KOTT375-00002 KOTT375-00003 KOTT375-00004 KOTT375-00005 KOTT375-00006 KOTT375-00007 KOTT375-00008 KOTT375-00009 KOTT375-00010 KOTT375-00011 KOTT375-00013
Chemical storage cabinet
Kalusteet Säilytyskaapit
These cabinets are designed for the storage of non-combustible hazardous materials.
  • Epoxy powder coated steel
  • Lockable doors
  • Optional integrated fan
  • Continuous extraction inside the cabinet
  • Cabinets without integrated fan should be connected to an extraction system
Maximum loading capacity: 30 kg per drawer.