Safety cabinets for flammable liquids, type 90
Toimittaja: KOETTERMANN
371-00024 371-00025 371-00026 371-00027 371-00028 371-00029 371-00030 371-00021 371-00008 371-00001 371-00002 371-00003 371-00004 371-00005 371-00023 371-00007 371-00022 371-00009 371-00010 371-00012 371-00015 371-00016 371-00017 371-00018 371-00019 371-00020 371-00006
KOTT371-00024EA 7580 EUR
KOTT371-00024 KOTT371-00025 KOTT371-00026 KOTT371-00027 KOTT371-00028 KOTT371-00029 KOTT371-00030 KOTT371-00021 KOTT371-00008 KOTT371-00001 KOTT371-00002 KOTT371-00003 KOTT371-00004 KOTT371-00005 KOTT371-00023 KOTT371-00007 KOTT371-00022 KOTT371-00009 KOTT371-00010 KOTT371-00012 KOTT371-00015 KOTT371-00016 KOTT371-00017 KOTT371-00018 KOTT371-00019 KOTT371-00020 KOTT371-00006
Safety cabinets for flammable liquids, type 90
Kalusteet Säilytyskaapit
Fire resistant up to 90 minutes safety cabinet with shelves for the storage of hazardous substances in working spaces. Incase of fire, the doors closes automatically, and the door frames are sealed by special fire safety seals.
  • Automatic door closure after one minute
  • Continuous extraction of fumes inside the cabinet
  • Lockable
  • Corrosion resistant door closures
Epoxy powder coated steel, thermal insulation at both interior and exterior.
Sertifikaatit: Classification type 90 in accordance with DIN EN 14470-1.