Wall-mounted cabinets
Toimittaja: KOETTERMANN
307-00052 307-00053 307-00054 307-00055 307-00056 307-00057 307-00058 307-00059 307-00060 307-00061 307-00050 307-00063 307-00049 307-00062 307-00036 307-00033 307-00051 307-00035 307-00037 307-00038 307-00039 307-00040 307-00041 307-00042 307-00043 307-00044 307-00045 307-00046 307-00047 307-00048 307-00034
KOTT307-00052EA 471 EUR
KOTT307-00052 KOTT307-00053 KOTT307-00054 KOTT307-00055 KOTT307-00056 KOTT307-00057 KOTT307-00058 KOTT307-00059 KOTT307-00060 KOTT307-00061 KOTT307-00050 KOTT307-00063 KOTT307-00049 KOTT307-00062 KOTT307-00036 KOTT307-00033 KOTT307-00051 KOTT307-00035 KOTT307-00037 KOTT307-00038 KOTT307-00039 KOTT307-00040 KOTT307-00041 KOTT307-00042 KOTT307-00043 KOTT307-00044 KOTT307-00045 KOTT307-00046 KOTT307-00047 KOTT307-00048 KOTT307-00034
Wall-mounted cabinets
Kalusteet Säilytyskaapit
Made of epoxy powder coated steel, with one shelf.
  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg