Storage Boxes for Vials/Bottles
Toimittaja: KOEHLER TECHNISCHE PRODUKTEN
2-7209
KOEH2-7209EA 8.5 EUR
KOEH2-7209
Storage Boxes for Vials/Bottles
Laatikot Laatikot vials
Storage boxes are suitable for bottles and vials which is used for cooling in freezers.
  • With trim insert and tight-closing lid
  • 50 places (5×10) for vessels with 12 mm Ø
Accepts autosampler bottle (1,2 ml), micro vial (1,5 ml), conic ampoule.