Storage Boxes
Toimittaja: KOEHLER TECHNISCHE PRODUKTEN
2-1907 2-1908 2-1909 2-1917 2-1918 2-1919 2-1920 2-1921 2-1922 2-1923 2-1924 2-1925
KOEH2-1907EA 18 EUR
KOEH2-1907 KOEH2-1908 KOEH2-1909 KOEH2-1917 KOEH2-1918 KOEH2-1919 KOEH2-1920 KOEH2-1921 KOEH2-1922 KOEH2-1923 KOEH2-1924 KOEH2-1925
Storage Boxes
Laatikot Laatikot putkille
Box in bright colours made of PP for cell culture tubes of 15 or 50 ml (Falcon). Two grids guarantee a safe stand of the tubes in the upper as well as the lower area.
  • Clear lid is providing the correct view
  • Temperature resistant from −80 °C up to +121 °C
  • Autoclavable