Woulff bottles, Erlenmeyer shape
Toimittaja: KNF
057953 057954
KNFA057953EA 0 EUR
KNFA057953 KNFA057954
Woulff bottles, Erlenmeyer shape
Pullot Woulff-pullot
Made of borosilicate glass 3.3. plastic coated.
  • Thick-walled Erlenmeyer flask with plastic safety coating
  • Vacuum gauge with two scales (1000 to 0 mbar, 760 to 0 mm Hg)
  • PVC ventilation valve for vacuum regulation
  • Glass port thread: GL 18 with plastic fittings for ³/₈" Int. Ø. tubing
  • GL 45 threaded bottle neck
Traps vapor/liquids to prolong pump life.