HPLC columns, Eurospher II, C18
Toimittaja: KNAUER
15VE181ESJ 25WE181ESJ 15EE181ESJ 25EE181E2J 15VE181E2J 25EE181ESJ 12DE181E2J
KNAU15VE181ESJEA 619 EUR
KNAU15VE181ESJ KNAU25WE181ESJ KNAU15EE181ESJ KNAU25EE181E2J KNAU15VE181E2J KNAU25EE181ESJ KNAU12DE181E2J
HPLC columns, Eurospher II, C18
Kromatografiakolonnit
These columns with HPLC phase are perfectly suited to take on routine analyses as well as the most ambitious chromatography tasks.
  • Spherical high performance HPLC phase based on silica gel
  • Unpolar, monomeric C18 (octadecyl) modification
The C18 phase can be considered for the determination of water soluble vitamins, steroids, catecholamines, sedatives and in many other application fields in the analytical as well as preparative range.