Embedding cassettes with disposable lid
Toimittaja: KLINIPATH
110-02T 110-06T
KLIN110-02TEA 165 EUR
KLIN110-02T KLIN110-06T
Embedding cassettes with disposable lid
Histologiset kasetit Kasetit kudosten käsittelyyn
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Tilaustiedot: Available in different colours.
Pakkaus: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.