Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Toimittaja: KIMBERLY CLARK
34256 34155
115-0285EA 0 EUR
115-0285 KIMB34155
Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Pyyhkeet, liinat ja sienet Kertakäyttöliinat ja -pyyhkeet Paperiliinat herkkiin kohteisiin
These single ply, light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.
  • Anti-stat dispensing reduces lint and electrostatic discharge and controls usage
  • Portability reduces waste
  • One-at-a-time dispensing (pop-up dispenser)