Urine container, with collection device for vacuum test tubes
Toimittaja: KIMA SAS DI CHIARIN RENZO
21800 21820 21780 21883 21870 21890 21870E
720-2862EA 688 EUR
720-2862 KIMA21820 720-2861 KIMA21883 KIMA21870 KIMA21890 KIMA21870E
Urine container, with collection device for vacuum test tubes
Diagnostiset testit Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Sterile containers for urine culture or non sterile for physical chemical microscopic analysis.
  • Collection spot with a partially removable adhesive label to be reclosed after the collection
  • The labels are printed with patient and operators instructions