Blood collection tubes, K3EDTA, EMOPLASTIK®
Toimittaja: KIMA SAS DI CHIARIN RENZO
18553
KIMA18553EA 0 EUR
KIMA18553
Blood collection tubes, K3EDTA, EMOPLASTIK®
Diagnostiset testit Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Polypropylene test tube with cap.
  • K3 EDTA for 5 ml of blood
  • Non sterile
  • With writing area