Safety blood collection holders with needle
Toimittaja: KIMA SAS DI CHIARIN RENZO
15521 15522 15566 15567
613-2964EA 50.9 EUR
613-2964 613-2965 KIMA15566 KIMA15567
Safety blood collection holders with needle
Neulat
Sterile holders with integrated and oriented needles for blood collection.
Available in different colours and gauges.
Shelf life: 36 months