Lids for reaction vessels
Toimittaja: KGW
D4278-S D4303-S D4304
KGWAD4278-SEA 0 EUR
KGWAD4278-S KGWAD4303-S KGWAD4304
Lids for reaction vessels
Kannet Reaction Vessel Lids
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes