Cold traps, long version, type KFL
Toimittaja: KGW
17380 17382 17381 17362
KGWA17380EA 1100 EUR
KGWA17380 KGWA17382 KGWA17381 KGWA17362
Cold traps, long version, type KFL
Kylmäloukut
These cold traps (cold finger condensers) are constructed in the same way as standard cold traps.
  • Available with or without spouts
  • Complete versions include Dewar flask and plastic ring
The condensation area of the cold traps has been extended in order to trap more condensation. In addition to this, the complete cold trap versions also have a larger plastic ring and a bigger Dewar flask.
An upgraded version is a long version cold trap with an outlet spout. These cold traps are constructed in the same way as standard cold traps but have a GL 32More Product Information