Dewar flasks with screw flange, Type GEW
Toimittaja: KGW
10911
KGWA10911EA 738 EUR
KGWA10911
Dewar flasks with screw flange, Type GEW
Nestetyppisäiliöt
Dewar flask made of borosilicate glass 3,3 with blue-coated metal cover.
  • Easy handling
  • With screw flange
Applications include protected from overflow, lockable and gassing coolants can be used but it should not exceed the maximum pressure of 0,1 bar.