Punnituslasi, matala, hiostulpalla
Toimittaja: Kavalier
632421202505 632421202343 632421202604 632421202605 632421202804 632421202503
KAVA632421202505EA 1140 EUR
KAVA632421202505 KAVA632421202343 KAVA632421202604 KAVA632421202605 KAVA632421202804 KAVA632421202503
Punnituslasi, matala, hiostulpalla
Pullot Punnituslasit
Made of borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance