Sentrifugiputket
Toimittaja: KARTELL
88304-03 88303/03 8830304 88303/06 8830310
KART88304-03EA 58.6 EUR
KART88304-03 KART88303/03 KART8830304 KART88303/06 KART8830310
Sentrifugiputket
Putket Sentrifugiputket
PP with quick closure PE screw cap.
  • Leak-proof
  • Can be centrifuged up to 4000×g
  • Cap is designed to avoid any contamination due to drops of liquid on the inner surface