Centrifuge tubes, conical bottom
Toimittaja: KARTELL
84008 84013 84006 84007 84011 84012 84009 84010 84015 84014
KART84008EA 540 EUR
KART84008 KART84013 KART84006 KART84007 KART84011 KART84012 KART84009 KART84010 KART84015 KART84014
Centrifuge tubes, conical bottom
Putket Sentrifugiputket
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Graduated
Conical bottom with tight screw cap.