Dishes, crystallising
Toimittaja: KARTELL
639 630 634
KART639EA 91.2 EUR
KART639 KART630 KART634
Dishes, crystallising
Haihdutusmaljat/Tarjottimet/Altaat Kiteytysmaljat
Crystallising dishes, made of PTFE.
  • Chemical, mechanical and heat resistance
  • Used safely for continuous service from −270 up to +250 °C
Suitable for dissolutions, digestions with acids and corrosive agents and suitable for food stuffs.