Uurretut survimet, PTFE-pää
Toimittaja: KARTELL
6315 6302 6305 6310 6330 6350
KART6315EA 120 EUR
KART6315 KART6302 KART6305 KART6310 KART6330 KART6350
Uurretut survimet, PTFE-pää
Huhmareet ja survimet Huhmareet
Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.
  • Head made of PTFE
  • Shaft made of stainless steel