Sleeves for vacuum filtration
Toimittaja: KARTELL
420
KART420EA 60.7 EUR
KART420
Sleeves for vacuum filtration
Pullotuet Tukihylsyt
Set of 7 adapters, conical, made of black neoprene, sleeve supports for Büchner funnels and filter flasks.