Petri dishes, disposable
Toimittaja: KARTELL
363 358 359 360 361 362 364 365 367 369 368 3360 380 0036600
KART363EA 458 EUR
KART363 KART358 KART359 KART360 KART361 KART362 KART364 KART365 KART367 KART369 KART368 KART3360 KART380 KART0036600
Petri dishes, disposable
Soluviljelyalustat Petrimaljat
Disposable planar, clear Petri dishes in PS. Available in several diameters, ventilated and unventilated, aseptic or radiation-sterilised. Styles with 2, 3 and 4 separate compartments. Designed for routine lab work and bacteriological research. Available with aeration notches on the lid to prevent condensation formation, or without aeration notches to avoid agar dessication after incubation.