Pipetti, Pasteur pipetti
Toimittaja: KARTELL
88337 334 88337/1
612-2815EA 0 EUR
612-2815 KART334 KART88337/1
Pipetti, Pasteur pipetti
Pipetit ja pipetintäyttäjät Pasteur-/siirtopipetit
PE, disposable, sterile or non sterile.
  • Disposable PE pipettes with bulb and graduations
  • The slender tip guarantees precision transfer of the sample
  • Non sterile pipettes are available in multi-packs; sterile pipettes are individually wrapped
Varoitus: *Capillary