Dekantterit, matala malli
Toimittaja: KARTELL
1534 1535 1536 1537 1538 1539
KART1534EA 87.6 EUR
KART1534 KART1535 KART1536 KART1537 KART1538 KART1539
Dekantterit, matala malli
Dekantterit
PFA, translucent, with spout.
  • Space-saving stackable design
  • High thermal resistance from −250 to +270 °C
  • Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.