Multiple or single cuvette segments
Toimittaja: KARTELL
1500 1501
KART1500EA 1020 EUR
KART1500 KART1501
Multiple or single cuvette segments
Kyvetit Erikoiskyvetit
These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Pakkaus: In cases of bulk segments (450) or 30 pre-racked segments (15).