Single channel pipettes, fixed volume, Fixopet
Toimittaja: KARTELL
1303.10 1304.10 1305.10 1306.10 1307.10 1308.10 1309.10 1310.10 1311.10 1312.10 1313.10
KART1303.10EA 426 EUR
KART1303.10 KART1304.10 KART1305.10 KART1306.10 KART1307.10 KART1308.10 KART1309.10 KART1310.10 KART1311.10 KART1312.10 KART1313.10
Single channel pipettes, fixed volume, Fixopet
Pipetit ja pipetintäyttäjät Pipetit
These single channel pipettes are available with fixed volumes in a range from 5 to 1000 µl.
  • Special handle ensures a more comfortable grip
  • Rotary device for the tip ejection does not create strain on the tendons of the thumb
  • Removable ejector: Can be used with test tubes with a very narrow opening
  • Integrated recalibration system ensures quick and precise recalibrations
  • Robust structure of the PVDF body ensures high mechanical and chemical resistances
  • Resistant and durable
  • Completely autoclavable at +121 °C for 20 minutes