Single channel pipettes, variable volume, Pluripet
Toimittaja: KARTELL
1296.10 1300.10 1301.10 1319.10 1321.10 1322.10 1325.10 1329.10
KART1296.10EA 856 EUR
KART1296.10 KART1300.10 KART1301.10 KART1319.10 KART1321.10 KART1322.10 KART1325.10 KART1329.10
Single channel pipettes, variable volume, Pluripet
Pipetit ja pipetintäyttäjät Pipetit
These variable pipettes are equipped with an accurate and precise pipetting mechanism. They cover the range from 0,2 to 10000 µl.
  • Special handle ensures a good grip and prevents hand fatigue
  • Volume can be selected by turning the emptying button or the cylinder
  • Height adjustable tip ejector houses a large variety of tips and can be disassembled to facilitate cleaning operations
  • Colour-coded buttons for easy identification
  • Robust PVDF body ensures high mechanical and chemical resistance
  • Completely autoclavable at +121 °C for 20 minutes
The PL2 and PL10More Product Information