Micro Test Tubes, Graduated
Toimittaja: KARTELL
0027904 0027906 0027912 0027913
KART0027904EA 398 EUR
KART0027904 KART0027906 KART0027912 KART0027913
Micro Test Tubes, Graduated
Putket Klusteriputket
Ideal for samples analysis.
  • Made from PP
  • Flat cap with writing area
  • Available in blue, yellow, pink and green
Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11000.
Sertifikaatit: The raw materials with the relevant requirements of Food-contact EU; Regulation 1935/2004 EC, Regulation 2023/2006/EC, FDA and REACH/ROHS regulations.