Disinfectant spray, Incidin® Liquid, Ecolab®
Toimittaja: KAMMERER
1612611 YLS12 1612621
KAMM1612611EA 0 EUR
KAMM1612611 KAMMYLS12 KAMM1612621
Disinfectant spray, Incidin® Liquid, Ecolab®
Puhdistusaineet Desinfiointiaineet
A disinfectant alcohol solution with broad spectrum of activity against bacteria (incl. TB) and yeasts. Reliable and rapid disinfection of all alcohol-resistant surfaces, even in high-risk areas.
  • Ready to use spray solution with particularly short reaction time and pleasant odour
  • Formaldehyde-free
  • Good wetting, rapid and residue-free drying
Varoitus: Use biocides safely. Always read the label and product information before use.