Stopper Bag Tubings, Tyvek® 1073B/HDPE
Toimittaja: KEYSTONE CLEANROOM PRODUCTS
SBTMGTVK02 SBTMGTVK03 SBTMGTVK04 SBTMGTVK06 SBTMGTVK09 SBTMGTVK10 SBTMGTVK12 SBTMGTVK18 SBTMGTVK28 018SBT12SD 028SBTWGC SBTMGTVK07 SBTMGTVK14 SBTMGTVK24 SBTMGTVK28SDON SBTMGTVK14SDON SBTMGTVK06SDON
129-3024EA 818 EUR
129-3024 129-3026 129-3027 129-3028 129-3029 129-3035 129-3036 129-3037 129-3038 KACC018SBT12SD KACC028SBTWGC KACCSBTMGTVK07 KACCSBTMGTVK14 KACCSBTMGTVK24 KACCSBTMGTVK28SDON KACCSBTMGTVK14SDON KACCSBTMGTVK06SDON
Stopper Bag Tubings, Tyvek® 1073B/HDPE
Pussit ja sulkijat Sterilointipussit
Tyvek® 1073B/HDPE autoclave continuous form tubing is ideal for steam sterilisation of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required. Tubing form allows for the creation of bags of any length, giving excellent sizing flexibility.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • One side made of hydrophobic and lint-free Tyvek® 1073B
  • One side made of semi-transparent HDPE film