Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Toimittaja: KEYSTONE CLEANROOM PRODUCTS
BWLKCRP30030 BWLKCRP30040 BWLKCRP30036 BWLKCRP30048 BWLKCRP30020 BWLKCRP30042 BWLKCRP30050 BWLKCRP30054 BWLKCRP30024 BWLKCRP30060
129-4013EA 5010 EUR
129-4013 129-4019 KACCBWLKCRP30036 KACCBWLKCRP30048 KACCBWLKCRP30020 KACCBWLKCRP30042 KACCBWLKCRP30050 KACCBWLKCRP30054 KACCBWLKCRP30024 KACCBWLKCRP30060
Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Kuoret
This 50 g/m² SMS autoclavable bowl cover provides a microbial barrier that is ideal for pharmaceutical production applications.
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Made in USA
Eliminate the need for cutting sheets of sterilisation wrapping materials and securing them with autoclave tape and rubber bands. After the outer bag is removed, thisMore Product Information