VWR®, Agarose I™
Toimittaja: VWR Chemicals
0710-100GEA 255 EUR
0710-100G 0710-25G 0710-500G K857-100TABS 0710-250G
VWR®, Agarose I™
Elektroforeesigeelit Agaroosi ja akryyliamidi
Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications (250 bp to 22 kb).
  • All-purpose, high purity agarose
  • Exceptional band resolution and clarity
  • Nuclease- and protease-free
Agarose I™ has a low EEO (electroendosmosis) for shorter electrophoretic runs without compromising resolution. Agarose I™ is excellent for blotting techniques and general manipulations.