VWR® BANDit™ Protein Stain Blue
Toimittaja: VWR Chemicals
K217-1LEA 271 EUR
K217-1L
VWR® BANDit™ Protein Stain Blue
Värit Elektroforeesivärit
Blue BANDit is a safe and environmentally friendly alternative to traditional Coomassie® Blue staining.
  • Reduces handling of hazardous chemicals: No methanol or acetic acid required
  • High sensitivity, quick, simple procedure